MAT184122952 Matrix Scientific

6,7,8,9-Tetrahydro-5H-carbazol-3-ylamine

  • MDL number MFCD07643278
  • Cas Number : 65796-52-3
  • Molecular Weight : 186.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033632.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033632.html
Availability details for
Quantity
  • Availability
  • Add to Cart