MAT184122988 Matrix Scientific

3-(2-Methoxy-phenyl)-4,5-dihydro-isoxazole-5-carboxylic acid

  • MDL number MFCD06011105
  • Cas Number : 508182-19-2
  • Molecular Weight : 221.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033668.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033668.html
Availability details for
Quantity
  • Availability
  • Add to Cart