MAT184123031 Matrix Scientific

(1-Aminomethyl-cyclohexyl)-dimethyl-amine

  • MDL number MFCD05201342
  • Cas Number : 41806-09-1
  • Molecular Weight : 156.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033711.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033711.html
Availability details for
Quantity
  • Availability
  • Add to Cart