MAT184123041 Matrix Scientific

1-Amino-3-(3-methyl-piperidin-1-yl)-propan-2-ol

  • MDL number MFCD00776902
  • Cas Number : 499975-59-6
  • Molecular Weight : 172.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033721.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033721.html
Availability details for
Quantity
  • Availability
  • Add to Cart