MAT184123077 Matrix Scientific

C-Phenyl-C-pyridin-3-yl-methylaminedihydrochloride

  • MDL number MFCD01603629
  • Cas Number : 287111-68-6
  • Molecular Weight : 257.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033757.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033757.html
Availability details for
Quantity
  • Availability
  • Add to Cart