MAT184123098 Matrix Scientific

4-Furan-2-yl-4-oxo-butyric acid

  • MDL number MFCD00092710
  • Cas Number : 10564-00-8
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033778.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033778.html
Availability details for
Quantity
  • Availability
  • Add to Cart