MAT184123131 Matrix Scientific

2-(5-Methyl-thiophen-2-yl)-ethylamine

  • MDL number MFCD08059759
  • Cas Number : 30433-92-2
  • Molecular Weight : 141.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033432.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033432.html
Availability details for
Quantity
  • Availability
  • Add to Cart