MAT184123174 Matrix Scientific

2-Chloro-5-pyrrol-1-yl-phenylamine

  • MDL number MFCD07396247
  • Cas Number : 878722-12-4
  • Molecular Weight : 192.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033854.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033854.html
Availability details for
Quantity
  • Availability
  • Add to Cart