MAT184123216 Matrix Scientific

[2-Amino-4-(4-fluoro-phenyl)-thiazol-5-yl]-acetic acid

  • MDL number MFCD08052633
  • Cas Number : 775519-63-6
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033896.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033896.html
Availability details for
Quantity
  • Availability
  • Add to Cart