MAT184123264 Matrix Scientific

4-Ethyl-4,5,6,7-tetrahydro-thieno[3,2-c]pyridinehydrochloride

  • MDL number MFCD07345579
  • Cas Number : 1052551-81-1
  • Molecular Weight : 203.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033944.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033944.html
Availability details for
Quantity
  • Availability
  • Add to Cart