MAT184123269 Matrix Scientific

3-Ethoxy-4-hydroxy-benzoic acid

  • MDL number MFCD06203134
  • Cas Number : 5438-38-0
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033949.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033949.html
Availability details for
Quantity
  • Availability
  • Add to Cart