MAT184123275 Matrix Scientific

3-(3,5-Dimethyl-1H-pyrazol-4-ylmethyl)-4-methoxy-benzaldehyde hydrochloride

  • MDL number MFCD11506558
  • Cas Number : 1033076-71-9
  • Molecular Weight : 280.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033955.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033955.html
Availability details for
Quantity
  • Availability
  • Add to Cart