MAT184123302 Matrix Scientific

4,5,6,7-Tetrahydro-benzo[b]thiophene-2-carbonyl chloride

  • MDL number MFCD04968788
  • Cas Number : 65361-26-4
  • Molecular Weight : 200.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033982.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033982.html
Availability details for
Quantity
  • Availability
  • Add to Cart