MAT184123312 Matrix Scientific

4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-thiazol-2-ylamine

  • MDL number MFCD04969875
  • Cas Number : 957301-85-8
  • Molecular Weight : 208.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033992.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033992.html
Availability details for
Quantity
  • Availability
  • Add to Cart