MAT184123358 Matrix Scientific

2,4-Dimethyl-6-piperazin-1-yl-pyrimidine

  • MDL number MFCD07365286
  • Cas Number : 887686-65-9
  • Molecular Weight : 192.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034038.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034038.html
Availability details for
Quantity
  • Availability
  • Add to Cart