MAT184123359 Matrix Scientific

3-(2,6-Dimethyl-pyrimidin-4-ylamino)-benzoic acid

  • MDL number MFCD07400491
  • Cas Number : 887686-71-7
  • Molecular Weight : 243.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034039.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034039.html
Availability details for
Quantity
  • Availability
  • Add to Cart