MAT184123362 Matrix Scientific

6-Acetylamino-nicotinic acid

  • MDL number MFCD00129118
  • Cas Number : 21550-48-1
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034042.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034042.html
Availability details for
Quantity
  • Availability
  • Add to Cart