MAT184123381 Matrix Scientific

1-Ethyl-2-methyl-2,3-dihydro-1H-indole-5-carbaldehyde

  • MDL number MFCD07790519
  • Cas Number : 887690-40-6
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034061.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034061.html
Availability details for
Quantity
  • Availability
  • Add to Cart