MAT184123399 Matrix Scientific

(2-Methyl-4-sulfamoyl-phenoxy)-acetic acid

  • MDL number MFCD07400616
  • Cas Number : 91004-91-0
  • Molecular Weight : 245.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034079.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034079.html
Availability details for
Quantity
  • Availability
  • Add to Cart