MAT184123403 Matrix Scientific

4-(1-Ethyl-2,5-dimethyl-1H-pyrrol-3-yl)-thiazol-2-ylamine

  • MDL number MFCD07400633
  • Cas Number : 887694-90-8
  • Molecular Weight : 221.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034083.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034083.html
Availability details for
Quantity
  • Availability
  • Add to Cart