MAT184123410 Matrix Scientific

(1-Furan-2-yl-propyl)-methyl-amine

  • MDL number MFCD07801195
  • Cas Number : 878441-50-0
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034090.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034090.html
Availability details for
Quantity
  • Availability
  • Add to Cart