MAT184123426 Matrix Scientific

5-(4-Chloro-phenyl)-2-ethyl-2H-pyrazole-3-carboxylic acid

  • MDL number MFCD08446012
  • Cas Number : 1015868-72-0
  • Molecular Weight : 250.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034106.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034106.html
Availability details for
Quantity
  • Availability
  • Add to Cart