MAT184123431 Matrix Scientific

1-(2-Methoxy-ethyl)-1H-benzoimidazole-5-carboxylic acid

  • MDL number MFCD07790838
  • Cas Number : 284672-82-8
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034111.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034111.html
Availability details for
Quantity
  • Availability
  • Add to Cart