MAT184123441 Matrix Scientific

2,5-Dimethyl-1-(7-methyl-2,3-dihydro-benzo[1,4]-dioxin-6-yl)-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD07402329
  • Cas Number : 879045-05-3
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034121.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034121.html
Availability details for
Quantity
  • Availability
  • Add to Cart