MAT184123446 Matrix Scientific

1-[5-Amino-1-(2-hydroxy-ethyl)-1H-benzoimidazol-2-yl]-ethanol dihydrochloride

  • MDL number MFCD11506572
  • Cas Number : 1185293-40-6
  • Molecular Weight : 294.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034126.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034126.html
Availability details for
Quantity
  • Availability
  • Add to Cart