MAT184123455 Matrix Scientific

(3-Morpholin-4-ylmethyl-indol-1-yl)-acetic acid

  • MDL number MFCD07402414
  • Cas Number : 879038-35-4
  • Molecular Weight : 274.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034135.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034135.html
Availability details for
Quantity
  • Availability
  • Add to Cart