MAT184123472 Matrix Scientific

5-Isopropyl-4-methoxy-2-methyl-benzylamine

  • MDL number MFCD07403241
  • Cas Number : 879063-02-2
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034152.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034152.html
Availability details for
Quantity
  • Availability
  • Add to Cart