MAT184123484 Matrix Scientific

2,5-Dimethyl-1-pyridin-2-ylmethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD07791605
  • Cas Number : 879053-20-0
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034164.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034164.html
Availability details for
Quantity
  • Availability
  • Add to Cart