MAT184123485 Matrix Scientific

3-Fluoro-4-pyrrolidin-1-yl-phenylaminedihydrochloride

  • MDL number MFCD08087469
  • Cas Number : 93246-54-9
  • Molecular Weight : 253.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034165.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034165.html
Availability details for
Quantity
  • Availability
  • Add to Cart