MAT184123499 Matrix Scientific

4-(2,4-Dimethyl-phenyl)-4-hydroxy-butyric acid

  • MDL number MFCD07792015
  • Cas Number : 879059-50-4
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034179.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034179.html
Availability details for
Quantity
  • Availability
  • Add to Cart