MAT184123580 Matrix Scientific

2,6-Dimethoxyaniline

  • MDL number MFCD00053934
  • Cas Number : 2734-70-5
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-78
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/034260.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/034260.html
Availability details for
Quantity
  • Availability
  • Add to Cart