MAT184123597 Matrix Scientific

3-Cyclopentyloxy-phenylamine

  • MDL number MFCD09036966
  • Cas Number : 653604-38-7
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034277.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034277.html
Availability details for
Quantity
  • Availability
  • Add to Cart