MAT184123610 Matrix Scientific

2-Chloro-6,7-dimethoxy-3-methyl-quinoline

  • MDL number MFCD08899105
  • Cas Number : 577967-81-8
  • Molecular Weight : 237.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034290.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034290.html
Availability details for
Quantity
  • Availability
  • Add to Cart