MAT184123616 Matrix Scientific

2-(3-Piperidin-1-yl-propoxy)-benzoic acid

  • MDL number MFCD09039913
  • Cas Number : 919036-82-1
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034296.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034296.html
Availability details for
Quantity
  • Availability
  • Add to Cart