MAT184123617 Matrix Scientific

2-(3-Morpholin-4-yl-propoxy)-benzoic acid

  • MDL number MFCD09039915
  • Cas Number : 179994-92-4
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034297.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034297.html
Availability details for
Quantity
  • Availability
  • Add to Cart