MAT184123624 Matrix Scientific

5-(Piperidine-1-sulfonyl)-2,3-dihydro-1H-indole

  • MDL number MFCD07687988
  • Cas Number : 874593-99-4
  • Molecular Weight : 266.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034304.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034304.html
Availability details for
Quantity
  • Availability
  • Add to Cart