MAT184123667 Matrix Scientific

2-(2-Fluoro-phenyl)-2-methyl-propionic acid

  • MDL number MFCD11036900
  • Cas Number : 870849-49-3
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034347.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034347.html
Availability details for
Quantity
  • Availability
  • Add to Cart