MAT184123685 Matrix Scientific

2-(2-Ethyl-imidazol-1-yl)-propionic acidhydrochloride

  • MDL number MFCD11506601
  • Cas Number : 1219346-32-3
  • Molecular Weight : 204.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034365.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034365.html
Availability details for
Quantity
  • Availability
  • Add to Cart