MAT184123701 Matrix Scientific

2-[(3-Amino-benzyl)-(2-hydroxy-ethyl)-amino]-ethanol

  • MDL number MFCD08690217
  • Cas Number : 919021-32-2
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034381.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034381.html
Availability details for
Quantity
  • Availability
  • Add to Cart