MAT184123723 Matrix Scientific

2-Ethyl-2-methyl-5-oxo-tetrahydro-furan-3-carboxylic acid

  • MDL number MFCD11052208
  • Cas Number : 38840-97-0
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146
Country of Origin USA
Product Link http://www.matrixscientific.com/034403.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034403.html
Availability details for
Quantity
  • Availability
  • Add to Cart