MAT184123837 Matrix Scientific

(4,5,6-Trimethyl-2-oxo-2H-pyrimidin-1-yl)-acetic acid hydrochloride

  • MDL number MFCD11506627
  • Cas Number : 1185294-96-5
  • Molecular Weight : 232.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034556.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034556.html
Availability details for
Quantity
  • Availability
  • Add to Cart