MAT184123841 Matrix Scientific

3-[Methyl-(3-trifluoromethyl-phenyl)-amino]-propionic acid hydrochloride

  • MDL number MFCD11506631
  • Cas Number : 1185101-15-8
  • Molecular Weight : 283.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034572.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034572.html
Availability details for
Quantity
  • Availability
  • Add to Cart