MAT184123844 Matrix Scientific

3-[(3-Fluoro-phenyl)-methyl-amino]-propionic acidhydrochloride

  • MDL number MFCD11506634
  • Cas Number : 1185293-67-7
  • Molecular Weight : 233.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034575.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034575.html
Availability details for
Quantity
  • Availability
  • Add to Cart