MAT184123855 Matrix Scientific

3-(2-Oxo-2H-pyridin-1-yl)-propionic acidhydrobromide

  • MDL number MFCD11506643
  • Cas Number : 878775-97-4
  • Molecular Weight : 248.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034665.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034665.html
Availability details for
Quantity
  • Availability
  • Add to Cart