MAT184123909 Matrix Scientific

1,7-Dimethyl-2,4-dioxo-1,2,3,4-tetrahydro-pyrido[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD08548408
  • Cas Number : 110181-69-6
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034503.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034503.html
Availability details for
Quantity
  • Availability
  • Add to Cart