MAT184123965 Matrix Scientific

3-Morpholin-4-yl-3-phenyl-propionic acid

  • MDL number MFCD09035612
  • Cas Number : 937652-66-9
  • Molecular Weight : 235.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034559.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034559.html
Availability details for
Quantity
  • Availability
  • Add to Cart