MAT184123980 Matrix Scientific

4-(4-Methyl-furazan-3-ylmethoxy)-benzoic acid

  • MDL number MFCD08436267
  • Cas Number : 889941-07-5
  • Molecular Weight : 234.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034437.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034437.html
Availability details for
Quantity
  • Availability
  • Add to Cart