MAT184123982 Matrix Scientific

3-[(4-Fluoro-phenyl)-methyl-amino]-propionic acid

  • MDL number MFCD09755076
  • Cas Number : 944886-90-2
  • Molecular Weight : 197.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034576.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034576.html
Availability details for
Quantity
  • Availability
  • Add to Cart