MAT184123998 Matrix Scientific

1-(5-Amino-2-methyl-phenyl)-pyrrolidin-2-one

  • MDL number MFCD09045534
  • Cas Number : 69131-44-8
  • Molecular Weight : 190.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034592.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034592.html
Availability details for
Quantity
  • Availability
  • Add to Cart