MAT184124002 Matrix Scientific

[1-(4-Fluoro-phenyl)-propyl]-methyl-amine

  • MDL number MFCD09043960
  • Cas Number : 926262-89-7
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034441.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034441.html
Availability details for
Quantity
  • Availability
  • Add to Cart